DL-2-Hydroxybutyric acid 98%

Supplier: Ambeed
Danger

Synonyms: 2-Hydroxybutyric acid, 2-Hydroxybutanoic acid

A1481678-1G A1481678-250MG A1481678-5G
76664-208EA 75.33 USD
76664-208 76664-210 76664-212
DL-2-Hydroxybutyric acid 98%
DL-2-Hydroxybutyric acid

Precaución: for Research Use Only

Formula: C₄H₈O₃
MW: 104.11 g/mol
MDL Number: MFCD00070502
CAS Number: 600-15-7
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR