Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate 95%

Supplier: Ambeed
Warning

A1481668-100MG A1481668-1G A1481668-250MG
76664-202EA 13.8 USD
76664-202 76664-204 76664-206
Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate 95%
Ethyl 1-(3-chlorophenyl)-1H-imidazole-5-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₁ClN₂O₂
MW: 250.6809 g/mol
CAS Number: 2110819-17-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR