4-Bromo-3-iodobenzotrifluoride 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-3-iodobenzotrifluoride, 1-Bromo-2-iodo-4-(trifluoromethyl)benzene

A1481660-100G A1481660-25G
76664-198EA 274.78 USD
76664-198 76664-200
4-Bromo-3-iodobenzotrifluoride 98+%
4-Bromo-3-iodobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃BrF₃I
MW: 350.9 g/mol
MDL Number: MFCD09800691
CAS Number: 640280-28-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR