5-Fluoro-2-nitrobenzotrifluoride 98+%

Supplier: Ambeed
Warning

A1481655-100G A1481655-25G
76664-192EA 34 USD
76664-192 76664-194 76664-196
5-Fluoro-2-nitrobenzotrifluoride 98+%
5-Fluoro-2-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MDL Number: MFCD00039741
CAS Number: 393-09-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR