2,6-Dibromo-4-(trifluoromethyl)aniline 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Amino-3,5-dibromo-α,α,α-trifluorotoluene
A1481654-25G
A1481654-5G
76664-188EA
51.99
USD
76664-188
76664-190
2,6-Dibromo-4-(trifluoromethyl)aniline 98+%
2,6-Dibromo-4-(trifluoromethyl)aniline
Precaución: for Research Use Only
Formula:
C₇H₄Br₂F₃N MW: 318.92 g/mol |
MDL Number:
MFCD00068181 CAS Number: 72678-19-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...