2-Chloro-3-(trifluoromethyl)aniline 98+%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-α,α,α-trifluoro-m-toluidine

A1481653-100G A1481653-25G A1481653-5G
76664-182EA 139 USD
76664-182 76664-184 76664-186
2-Chloro-3-(trifluoromethyl)aniline 98+%
2-Chloro-3-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₅ClF₃N
MW: 195.57 g/mol
MDL Number: MFCD04115988
CAS Number: 62476-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR