4-Bromo-3-nitrotoluene 98+%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-4-methyl-2-nitrobenzene

A1481648-25G A1481648-5G A1481648-100G A1481648-500G
76664-162EA 15 USD
76664-162 76664-164 77597-516 77624-346
4-Bromo-3-nitrotoluene 98+%
4-Bromo-3-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 35
MDL Number: MFCD00024180
CAS Number: 5326-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR