2,5-Dibromo-m-xylene 98+%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dibromo-1,3-dimethylbenzene, 1,4-Dibromo-2,6-dimethylbenzene

A1481647-25G A1481647-5G
76664-158EA 35 USD
76664-158 76664-160
2,5-Dibromo-m-xylene 98+%
2,5-Dibromo-m-xylene

Precaución: for Research Use Only

Formula: C₈H₈Br₂
MW: 263.96
MDL Number: MFCD00079745
CAS Number: 100189-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR