2-Bromo-4,6-xylidine 98+%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2,4-dimethylaniline, 6-Bromo-2,4-xylidine, 2-Bromo-4,6-dimethylaniline

A1481646-100G A1481646-25G A1481646-5G
76664-152EA 158.08 USD
76664-152 76664-154 76664-156
2-Bromo-4,6-xylidine 98+%
2-Bromo-4,6-xylidine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
MDL Number: MFCD00047826
CAS Number: 41825-73-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR