2-Bromo-5-methylphenol ≥98%

Supplier: Ambeed
Warning

A1481645-5G A1481645-25G
76664-150EA 40.32 USD
76664-150 77592-784
2-Bromo-5-methylphenol ≥98%
2-Bromo-5-methylphenol

Precaución: for Research Use Only

Formula: C₇H₇BrO
MW: 187.034 g/mol
MDL Number: MFCD11100989
CAS Number: 14847-51-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR