1,4-Difluoro-2-methyl-5-nitrobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Difluoro-4-nitrotoluene

A1481642-100G A1481642-1G A1481642-25G A1481642-5G
76664-134EA 190 USD
76664-134 76664-136 76664-138 76664-140
1,4-Difluoro-2-methyl-5-nitrobenzene 98%
1,4-Difluoro-2-methyl-5-nitrobenzene

Precaución: for Research Use Only

Formula: C₇H₅F₂NO₂
MDL Number: MFCD06200905
CAS Number: 141412-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR