3-Fluoro-4-iodotoluene 98%

Supplier: Ambeed
Warning

A1481638-100G A1481638-25G A1481638-5G
76664-128EA 95 USD
76664-128 76664-130 76664-132
3-Fluoro-4-iodotoluene 98%
3-Fluoro-4-iodotoluene

Precaución: for Research Use Only

Formula: C₇H₆FI
MDL Number: MFCD00143162
CAS Number: 452-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR