(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 96%

Supplier: Ambeed
Warning

Synonyms: (R)-2-N-Boc-propane-1,2-diamine, tert-Butyl (R)-1-aminopropan-2-ylcarbamate

A1481568-250MG A1481568-1G A1481568-5G
76664-114EA 10 USD
76664-114 77592-780 77875-254
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate 96%
(R)-tert-Butyl (1-aminopropan-2-yl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD11112229
CAS Number: 100927-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR