Tetramethylammonium hydrogencarbonate 60% (w/w) in water

Supplier: Ambeed
Warning

A1481550-100G A1481550-25G
76664-110EA 28 USD
76664-110 76664-112
Tetramethylammonium hydrogencarbonate 60% (w/w) in water
Tetramethylammonium hydrogencarbonate

Precaución: for Research Use Only

Formula: C₅H₁₃NO₃
MDL Number: MFCD02684366
CAS Number: 58345-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR