2,6-Diisopropyl-phenyl-acenaphthoimidazolylidene-N-heterocyclic carbene-palladium chloride-benzyldimethylamine complex 95%

Supplier: Ambeed
Warning

A1481437-100MG A1481437-250MG
76664-104EA 183.54 USD
76664-104 76664-106
2,6-Diisopropyl-phenyl-acenaphthoimidazolylidene-N-heterocyclic carbene-palladium chloride-benzyldimethylamine complex 95%
2,6-Diisopropyl-phenyl-acenaphthoimidazolylidene-N-heterocyclic carbene-palladium chloride-benzyldimethylamine complex

Precaución: for Research Use Only

Formula: C₄₆H₅₂ClN₃Pd
MW: 788.7982 g/mol
CAS Number: 2757083-02-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR