4,6-Diaminopyrimidine 98%

Supplier: Ambeed
Warning

Synonyms: 4,6-Pyrimidinediamine

A1481407-1G A1481407-5G
76664-098EA 6.37 USD
76664-098 76664-100
4,6-Diaminopyrimidine 98%
4,6-Diaminopyrimidine

Precaución: for Research Use Only

Formula: C₄H₆N₄
MW: 110.12 g/mol
MDL Number: MFCD00044572
CAS Number: 2434-56-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR