5-(tert-Butoxycarbonylamino)-1-pentanol 98%

Supplier: Ambeed
Warning

Synonyms: N-(5-Hydroxypentyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(5-Hydroxypentyl)carbamate, tert-Butyl (5-hydroxypentyl)carbamate, N-Boc-5-aminopentanol, 5-(Boc-amino)-1-pentanol

A1481405-1G A1481405-250MG A1481405-5G A1481405-25G A1481405-100G
76664-092EA 18 USD
76664-092 76664-094 76664-096 77567-056 77624-342
5-(tert-Butoxycarbonylamino)-1-pentanol 98%
5-(tert-Butoxycarbonylamino)-1-pentanol

Precaución: For research use only.

Formula: C₁₀H₂₁NO₃
MDL Number: MFCD01862955
CAS Number: 75178-90-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR