1,3-Bis((3aS,8aR)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)benzene ≥97%, ee 99%

Supplier: Ambeed
Warning

A1481274-100MG A1481274-1G A1481274-250MG
76664-080EA 60.48 USD
76664-080 76664-082 76664-084
1,3-Bis((3aS,8aR)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)benzene ≥97%, ee 99%
1,3-Bis((3aS,8aR)-8,8a-dihydro-3aH-indeno[1,2-d]oxazol-2-yl)benzene

Precaución: for Research Use Only

Formula: C₂₆H₂₀N₂O₂
MW: 392.4492 g/mol
CAS Number: 175733-74-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR