4'-(N-(1-(Cyclopropanecarbonyl)-1,2,3,4-tetrahydroquinolin-7-yl)sulfamoyl)-[1,1'-biphenyl]-3-carboxamide 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
SU0268
A1481146-100MG
A1481146-50MG
76664-044EA
193.09
USD
76664-044
76664-046
4'-(N-(1-(Cyclopropanecarbonyl)-1,2,3,4-tetrahydroquinolin-7-yl)sulfamoyl)-[1,1'-biphenyl]-3-carboxamide 98+%
4'-(N-(1-(Cyclopropanecarbonyl)-1,2,3,4-tetrahydroquinolin-7-yl)sulfamoyl)-[1,1'-biphenyl]-3-carboxamide
Precaución: for Research Use Only
Formula:
C₂₆H₂₅N₃O₄S MW: 475.56 g/mol |
MDL Number:
MFCD34474075 CAS Number: 2210228-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...