Benzyl 6-oxo-5-oxa-2,7-diazaspiro[3.4]octane-2-carboxylate 97%

Supplier: Ambeed
Warning

A1481098-100MG A1481098-1G A1481098-250MG A1481098-50MG
76664-020EA 51 USD
76664-020 76664-022 76664-024 76664-026
Benzyl 6-oxo-5-oxa-2,7-diazaspiro[3.4]octane-2-carboxylate 97%
Benzyl 6-oxo-5-oxa-2,7-diazaspiro[3.4]octane-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₄N₂O₄
MDL Number: MFCD32735987
CAS Number: 1935064-61-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR