5-Fluorobenzofuran-6-carbonitrile 95%

Supplier: Ambeed
Warning

A1480464-100MG A1480464-1G A1480464-250MG
76664-014EA 25.47 USD
76664-014 76664-016 76664-018
5-Fluorobenzofuran-6-carbonitrile 95%
5-Fluorobenzofuran-6-carbonitrile

Precaución: for Research Use Only

Formula: C₉H₄FNO
MW: 161.1326 g/mol
MDL Number: MFCD23705168
CAS Number: 1427324-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR