(3aS,8aS)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 97%
Supplier: Ambeed
|
Warning
|
A1480140-1G
A1480140-250MG
76664-010EA
296
USD
76664-010
76664-012
(3aS,8aS)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine 97%
(3aS,8aS)-4,4,8,8-Tetrakis(3,5-diethylphenyl)-2,2-dimethyl-6-phenyltetrahydro-[1,3]dioxolo[4,5-e][1,3,2]dioxaphosphepine
Precaución: for Research Use Only
Formula:
C₅₃H₆₅O₄P MW: 797.0546 g/mol |
CAS Number:
2639939-72-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...