(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98%, ee 99%

Supplier: Ambeed
Warning

A1480106-1G A1480106-250MG A1480106-5G
76664-004EA 46 USD
76664-004 76664-006 77592-776
(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone ≥98%, ee 99%
(S)-(-)-4-Isopropyl-5,5-diphenyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₈H₁₉NO₂
MDL Number: MFCD03093556
CAS Number: 184346-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR