5-(Dimethylphosphoryl)furan-2-carboxylic acid 95%

Supplier: Ambeed
Danger

A1479865-100MG A1479865-250MG
76664-002EA 127 USD
76664-002 77875-244
5-(Dimethylphosphoryl)furan-2-carboxylic acid 95%
5-(Dimethylphosphoryl)furan-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₉O₄P
CAS Number: 2361984-11-2
UN: 3464
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR