1-Methyl-6-(methylamino)pyrimidine-2,4(1H,3H)-dione 97%
Supplier: Ambeed
|
Warning
|
A1479614-100MG
A1479614-250MG
76663-998EA
51.99
USD
76663-998
76664-000
1-Methyl-6-(methylamino)pyrimidine-2,4(1H,3H)-dione 97%
1-Methyl-6-(methylamino)pyrimidine-2,4(1H,3H)-dione
Precaución: for Research Use Only
Formula:
C₆H₉N₃O₂ MW: 155.1546 g/mol |
MDL Number:
MFCD20665015 CAS Number: 89465-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...