(1R,2R)-N1,N2-Dimethyl-1,2-bis(4-(trifluoromethyl)phenyl)ethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A1477242-100MG A1477242-1G A1477242-250MG
76663-978EA 98.67 USD
76663-978 76663-980 76663-982
(1R,2R)-N1,N2-Dimethyl-1,2-bis(4-(trifluoromethyl)phenyl)ethane-1,2-diamine 97%
(1R,2R)-N1,N2-Dimethyl-1,2-bis(4-(trifluoromethyl)phenyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₈H₁₈F₆N₂
MW: 376.3394 g/mol
CAS Number: 220665-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR