3-Iodo-o-toluic acid 96%

Supplier: Ambeed
Warning

A1477236-25G A1477236-5G
76663-974EA 51.99 USD
76663-974 76663-976
3-Iodo-o-toluic acid 96%
3-Iodo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MW: 262.05 g/mol
MDL Number: MFCD00270112
CAS Number: 133232-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR