3-Bromo-2-chloroanisole 98+%

Supplier: Ambeed
Warning

A1477224-1G A1477224-25G A1477224-5G
76663-966EA 48 USD
76663-966 76663-968 76663-970
3-Bromo-2-chloroanisole 98+%
3-Bromo-2-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MDL Number: MFCD16036127
CAS Number: 174913-11-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR