2-Bromo-1-(o-tolyl)ethanone 97%

Supplier: Ambeed
Danger

A1477218-1G A1477218-25G A1477218-5G
76663-952EA 26 USD
76663-952 76663-954 76663-956
2-Bromo-1-(o-tolyl)ethanone 97%
2-Bromo-1-(o-tolyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₉BrO
MDL Number: MFCD04038955
CAS Number: 51012-65-8
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR