4,4',4''-((4,4',4''-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(methylene))tribenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A1477134-50MG
A1477134-100MG
A1477134-1G
A1477134-250MG
76663-948EA
41
USD
76663-948
77875-227
77875-228
77875-229
4,4',4''-((4,4',4''-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(methylene))tribenzoic acid 97%
4,4',4''-((4,4',4''-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(methylene))tribenzoic acid
Precaución: for Research Use Only
Formula:
C₃₃H₃₀N₁₀O₆ |
MDL Number:
MFCD23380185 CAS Number: 1093949-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...