(2-(2-Hydroxyethyl)phenyl)boronic acid 97%

Supplier: Ambeed
Warning

A1477114-1G A1477114-250MG
76663-944EA 359.65 USD
76663-944 76663-946
(2-(2-Hydroxyethyl)phenyl)boronic acid 97%
(2-(2-Hydroxyethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₁BO₃
MW: 165.98 g/mol
MDL Number: MFCD18383604
CAS Number: 1232424-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR