6-Bromo-2,3-difluorobenzaldehyde 98+%

Supplier: Ambeed
Warning

A1477102-100G A1477102-25G A1477102-5G
76663-926EA 371.32 USD
76663-926 76663-928 76663-930
6-Bromo-2,3-difluorobenzaldehyde 98+%
6-Bromo-2,3-difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O
MDL Number: MFCD07368119
CAS Number: 360576-04-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR