2-Chloro-3-(trifluoromethyl)phenylboronic acid 98+%

Supplier: Ambeed
Warning

A1477096-1G A1477096-25G A1477096-5G
76663-920EA 13.8 USD
76663-920 76663-922 76663-924
2-Chloro-3-(trifluoromethyl)phenylboronic acid 98+%
2-Chloro-3-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₅BClF₃O₂
MW: 224.3726 g/mol
MDL Number: MFCD09258745
CAS Number: 957061-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR