4-Fluoro-2-formylphenylboronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoro-2-formylbenzeneboronic acid, (4-Fluoro-2-formylphenyl)boronic acid

A1477094-1G A1477094-25G A1477094-5G
76663-914EA 14.86 USD
76663-914 76663-916 76663-918
4-Fluoro-2-formylphenylboronic acid 98+%
4-Fluoro-2-formylphenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₆BFO₃
MW: 167.93 g/mol
MDL Number: MFCD10697426
CAS Number: 825644-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR