3,5-Difluoro-4-hydroxyphenylboronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (3,5-Difluoro-4-hydroxyphenyl)boronic acid

A1477092-1G A1477092-100MG
76663-912EA 244.01 USD
76663-912 77597-510
3,5-Difluoro-4-hydroxyphenylboronic acid 98+%
3,5-Difluoro-4-hydroxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD17011981
CAS Number: 1132666-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR