2,5-Dibromophenylboronic acid 98%

Supplier: Ambeed
Warning

A1477086-1G A1477086-25G A1477086-5G
76663-906EA 18.04 USD
76663-906 76663-908 76663-910
2,5-Dibromophenylboronic acid 98%
2,5-Dibromophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBr₂O₂
MDL Number: MFCD09743927
CAS Number: 1008106-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR