3-Bromo-2-fluorophenylboronic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-2-fluorophenyl)boronic acid, 3-Bromo-2-fluorobenzeneboronic acid

A1477082-100G A1477082-1G A1477082-25G A1477082-5G
76663-898EA 604.72 USD
76663-898 76663-900 76663-902 76663-904
3-Bromo-2-fluorophenylboronic acid 98+%
3-Bromo-2-fluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.816 g/mol
MDL Number: MFCD05664228
CAS Number: 352535-97-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR