2-Chloropyrimidine-5-boronic acid pinacol ester 98+%

Supplier: Ambeed
Warning

Synonyms: 2-chloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine, 2-chloro-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

A1477072-100G A1477072-1G A1477072-25G A1477072-5G
76663-882EA 577.13 USD
76663-882 76663-884 76663-886 76663-888
2-Chloropyrimidine-5-boronic acid pinacol ester 98+%
2-Chloropyrimidine-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₀H₁₄BClN₂O₂
MW: 240.494 g/mol
MDL Number: MFCD11856048
CAS Number: 1003845-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR