4-Chloro-3-nitrobenzoic acid 98+%

Supplier: Ambeed
Warning

A1477068-1000G A1477068-100G A1477068-25G A1477068-500G
76663-874EA 59 USD
76663-874 76663-876 76663-878 76663-880
4-Chloro-3-nitrobenzoic acid 98+%
4-Chloro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄ClNO₄
MDL Number: MFCD00007079
CAS Number: 96-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR