1-Nitro-9,10-anthraquinone 99+%

Supplier: Ambeed
Warning

A1477064-1000G A1477064-100G A1477064-25G A1477064-500G
76663-866EA 286 USD
76663-866 76663-868 76663-870 76663-872
1-Nitro-9,10-anthraquinone 99+%
1-Nitro-9,10-anthraquinone

Precaución: for Research Use Only

Formula: C₁₄H₇NO₄
MDL Number: MFCD00019140
CAS Number: 82-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR