Terephthalaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: Terephthaldialdehyde, Terephthaldicarboxaldehyde, p-Phthalaldehyde

A1477058-1000G A1477058-100G A1477058-25G A1477058-500G
76663-852EA 55.17 USD
76663-852 76663-854 76663-856 76663-858
Terephthalaldehyde 98%
Terephthalaldehyde

Precaución: for Research Use Only

Formula: C₈H₆O₂
MW: 134.13 g/mol
MDL Number: MFCD00006949
CAS Number: 623-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR