(2,4-Dichlorophenyl)acetic acid 98+%

Supplier: Ambeed
Warning

Synonyms: DCAA, 2,4-Dichlorophenylacetic acid

A1477052-1000G A1477052-100G A1477052-25G A1477052-500G
76663-836EA 126.25 USD
76663-836 76663-838 76663-840 76663-842
(2,4-Dichlorophenyl)acetic acid 98+%
(2,4-Dichlorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃Cl₂)CH₂COOH
MW: 205.04 g/mol
MDL Number: MFCD00004318
CAS Number: 19719-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR