2-(Ethoxycarbonyl)-6-nitrobenzoic acid 98+%

Supplier: Ambeed
Warning

A1477048-100G A1477048-25G A1477048-500G A1477048-5G
76663-828EA 46 USD
76663-828 76663-830 76663-832 76663-834
2-(Ethoxycarbonyl)-6-nitrobenzoic acid 98+%
2-(Ethoxycarbonyl)-6-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₆
MDL Number: MFCD01365751
CAS Number: 16533-45-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR