8-Amino-1-naphthalenesulfonic acid ≥98% (by HPLC) (contains < 20% H₂O)

Supplier: Ambeed
Warning

Synonyms: 8-Aminonaphthalene-1-sulphonic acid

A1476966-100G A1476966-25G A1476966-500G
76663-814EA 21.22 USD
76663-814 76663-816 76663-818
8-Amino-1-naphthalenesulfonic acid ≥98% (by HPLC) (contains < 20% H₂O)
8-Amino-1-naphthalenesulphonic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₃S
MW: 223.25 g/mol
MDL Number: MFCD00035730
CAS Number: 82-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR