1,3-Bis(4-hydroxybutyl)tetramethyldisiloxane 90% (by NMR)

Supplier: Ambeed
Warning

A1476898-1G A1476898-5G
76663-810EA 83 USD
76663-810 76663-812
1,3-Bis(4-hydroxybutyl)tetramethyldisiloxane 90% (by NMR)
1,3-Bis(4-hydroxybutyl)tetramethyldisiloxane

Precaución: for Research Use Only

Formula: C₁₂H₃₀O₃Si₂
MDL Number: MFCD00039554
CAS Number: 5931-17-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR