(E)-N-Benzylcinnamamide 95%

Supplier: Ambeed
Warning

A1476892-25G A1476892-5G
76663-806EA 305 USD
76663-806 76663-808
(E)-N-Benzylcinnamamide 95%
(E)-N-Benzylcinnamamide

Precaución: for Research Use Only

Formula: C₁₆H₁₅NO
MW: 237.2964 g/mol
CAS Number: 5100-00-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR