Acid Red 18 (Food Red No. 102) 85% (dye content)
Supplier: Ambeed
|
Warning
|
Synonyms:
Cochineal Red A, New Coccine, Scarlet 3R, Sodium 7-hydroxy-8-((4-sulphonatonaphthalen-1-yl)diazenyl)naphthalene-1,3-disulphonate, Ponceau 4R, Cochenille red
A1476867-100G
A1476867-25G
76663-804EA
45
USD
76663-804
77624-328
Acid Red 18 (Food Red No. 102) 85% (dye content)
Acid Red 18 (Food Red No. 102)
Precaución: for Research Use Only
Formula:
C₂₀H₁₄N₂O₁₀S₃ |
MDL Number:
MFCD00004084 CAS Number: 2611-82-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...