(4-(Decyloxy)phenyl)(phenyl)methanone 98+%

Supplier: Ambeed
Warning

A1476676-100G A1476676-25G A1476676-500G
76663-786EA 25.47 USD
76663-786 76663-788 76663-790
(4-(Decyloxy)phenyl)(phenyl)methanone 98+%
(4-(Decyloxy)phenyl)(phenyl)methanone

Precaución: for Research Use Only

Formula: C₂₃H₃₀O₂
MW: 338.4831 g/mol
CAS Number: 78392-97-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR