N-(3-(4-Fluorophenylsulfonamido)-4-hydroxyphenyl)-[1,1'-biphenyl]-4-carboxamide 98+%

Supplier: Ambeed
Warning

A1476412-100MG A1476412-25MG A1476412-50MG
76663-772EA 628.06 USD
76663-772 76663-774 76663-776
N-(3-(4-Fluorophenylsulfonamido)-4-hydroxyphenyl)-[1,1'-biphenyl]-4-carboxamide 98+%
N-(3-(4-Fluorophenylsulfonamido)-4-hydroxyphenyl)-[1,1'-biphenyl]-4-carboxamide

Precaución: for Research Use Only

Formula: C₂₅H₁₉FN₂O₄S
MW: 462.49 g/mol
CAS Number: 2249435-90-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR