3-Ethyl-1-vinyl-1H-imidazol-3-ium bromide 99+%

Supplier: Ambeed
Warning

A1476278-100G A1476278-25G A1476278-500G A1476278-5G
76663-758EA 114.58 USD
76663-758 76663-760 76663-762 76663-764
3-Ethyl-1-vinyl-1H-imidazol-3-ium bromide 99+%
3-Ethyl-1-vinyl-1H-imidazol-3-ium bromide

Precaución: for Research Use Only

Formula: C₇H₁₁BrN₂
MW: 203.0796 g/mol
CAS Number: 34311-88-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR